To run an analysis with the Workflow Editor, you must open the editor interface from within the study.
Procedure
- Follow either of the procedures in the following table.
Task | Procedure |
|---|---|
Open the Workflow Editor from inside an analysis | Procedure
|
Close the Workflow Editor inside an analysis | Select Close (X) on the Analysis title bar. NOTE Save your workflows before closing the analysis. There is no prompt to save. If you want to use a workflow in a different study, you must save the workflow and rename it. |