You can reprocess a result file by adding a new consensus workflow.
Prerequisites
  1. You have a study open.
Procedure
  1. Select the Analysis Results tab.
  2. Hold down the CTRL or SHIFT keys to select one or more analysis that you want to reprocess with a new consensus workflow.
  3. Select Reprocess, and then select Use Results to Create New (Multi) Consensus.
  4. The application adds the analysis associated with the selected result file.
  5. (Optional) Select any other analysis result that you want to reprocess with a new consensus workflow, select Reprocess, and select Use Results to Create New (Multi) Consensus.
  6. Select the Workflows tab, and then select the Consensus tab if it is not already open.
  7. Select or create a new consensus workflow. For instructions, see Workflows overview.
  8. The template opens in the Workflow Tree pane of the Workflow Editor.
  9. Select Run.