You can change the workflow, the settings, the parameters, and so on in an existing analysis before reprocessing.
Prerequisites
  1. You have a study open.
Procedure
  1. In an open study, select the Analysis Results tab if it is not already selected.
  2. Select the search that you want to reprocess.
  3. Select Reprocess.
  4. Select All Analysis Steps.
  5. Change the appropriate workflows, settings, parameters, input files, and so on.
  6. In the Analysis window, select Run.
  7. The job queue opens, displaying the status of your search. The application reruns only the nodes that are impacted by your changes. For information about the job queue, see Job queue.