You can remove an individual study name or a result file name from the Start Page.
Procedure
- Right-click the study name or result file that you want to delete.
- Select Remove From List.
NOTE
If you attempt to open a study or result file that you have deleted in Windows Explorer, the application prompts you to delete it from the Start Page.