You can copy and paste the text of the quatification method into Excel.
Procedure
  1. Select Administration > Maintain Quantification Methods.
  2. Select the pencil icon next to the method, and press CTRL+C.
  3. Paste the method into an Excel file.