When you select the Add Fractions button, the application treats all fractions as one sample but only applies appropriate chromatographic alignment and peak identification across the physical-property-matched fractions of the study.
Procedure
  1. In the study, select the Input Files tab.
  2. Select Add Fractions.
  3. In the Add Fractions dialog box, select the files that correspond to the fractionated sample.
  4. Select Open.
  5. The Proteome Discoverer application adds all selected files as a single study file.