Numeric study factors are used for time points, sample injection amounts, and so on. Numeric study factors sort numerically.
Procedure
- In the Study Factors area of the Study Definition page, select Add > Numerical Factor.
- The numerical factor dialog box opens, and its title bar includes the default name of [new factor].
- Select [new factor] and enter a name for the new numerical factor.
- In this example, the value is Time (days)
- In the box to the left of the Add and Delete buttons, enter the name of the first value, and then select Add.
- In this example, the value is 5.
- Continue adding additional values.
- Select Apply.
- The Study Definition page resembles the following image.
- (Optional) Repeat the steps above to add more factors.