You can deactivate a quantification method.
Procedure
  1. Do one of the following:
  2. Select Administration > Maintain Quantification Methods.
  3. Select Maintain Quantification Methods .
  4. The Quantification Methods view opens.
  5. Clear the checkbox in the Is Active column on the line containing the quantification method that you want to make inactive.
  6. (Optional) To make the quantification method active again, select the checkbox in the Is Active column on the line containing the quantification method that you want to make active.