You can add a new channel rather than modify an existing channel template, follow these instructions when the Quantification Method Editor opens.
Prerequisites
- You have the Quantification Method Editor dialog box open. See Create a quantification method.
Procedure
- Select the plus button (+) beneath the list of labels in the box on the far left.
- The following default name appears in the list of quantification channels in the box at the far left of the dialog box and in the Channel Name box: New Number.
- (Optional) To remove a quantification channel, select the quantification channel in the list of quantification channels and select the minus button (–) beneath the list.
- To specify a name for the new quantification channel, do the following:
- Delete the default name in the Channel Name box.
- Enter the new name.
- The new name now appears in the quantification channel (far left) box.
- To specify a quantification label to assign to the new quantification channel, select the plus button (+) beneath the Quantification Labels box.
- A default quantification label of New number now appears in the Quantification Labels box and the Label Name box.
- (Optional) To remove an existing quantification label, do the following:
- Select the label in the Quantification Labels box and select the minus button (–) beneath the box.
- Delete the label name in the Label Name box and then enter the new name.
- In the Modification dropdown box, select the modification for the amino acid label.
- In the box to the right of the Modification box, select the abbreviation of the amino acid selected in the Quantification Labels box that the modification should occur on.
- Select OK.
- The Save Quantification Method dialog box opens.
- In the Save as New Method box, enter the name of the new method.
- Select Save.
- The new quantification method appears in the Quantification Methods view.