If you have altered one of the quantification method templates listed at the beginning of Create a quantification method, you can restore the original template that was in effect when the application was first installed.
Procedure
- Do one of the following:
- Select Administration > Maintain Quantification Methods.
- Select Maintain Quantification Methods .
- The Quantification Methods view opens.
- To open the Quantification Method Editor dialog box, select Add in the Quantification Methods view.
- The Create a New Quantification Method dialog box opens.
- Select the appropriate template from the Create from Factory Defaults list.
- Set up the quantification method according to the instructions in Create a quantification method.