You can save a quantification method to use on another computer.
Procedure
- Do one of the following:
- Select Administration > Maintain Quantification Methods.
- Select Maintain Quantification Methods .
- The Quantification Methods view opens.
- Select the method that you want to export in the Quantification Methods view by selecting the column to the left of the Status column.
- Select Export.
- In the Export Quan Method dialog box, locate the folder where you want to store the METHOD file containing the quantification method, and select Save.
- The application saves the method in a file with the name of the method and a .method suffix, for example, TMT 2plex.method.