You can import a new quantification method from another computer.
Procedure
  1. Do one of the following:
  2. Select Administration > Maintain Quantification Methods.
  3. Select Maintain Quantification Methods .
  4. The Quantification Methods view opens.
  5. Select Import.
  6. In the Import Quan Method dialog box, select the METHOD file containing the method that you want to import, and select Open.
  7. If the new method is valid, the Quantification Method Editor dialog box opens, showing the new method.
  8. If the new method is not valid, a message box opens that describes the error.
  9. For a valid method, select OK in the Quantification Method Editor dialog box.
  10. Change the name of the imported quantification method by changing it in the Method Name column of the Quantification Methods view.