Procedure
- In the Annotation Aspect view on the Administration page, select Add in the toolbar.
- The Annotation Aspect Editor opens.
- The Annotation Aspects Editor consists of three parts:
- On the top is the aspect definition area, which includes the name and description of the annotation aspect and colored boxes representing the annotation groups of the aspect.
- On the lower left is the Annotation Group area, which contains the definitions of a single annotation group.
- On the lower right is the Available Annotations area, which lists the available annotations.
- In the Aspect Name box in the aspect definition area, enter the aspect name.
NOTE
Spaces are not supported. You can use letters, numbers, and underscores (_).
- In the Description box, enter a brief definition of the new aspect.
- Select New Group in the Annotation Group area of the Editor.
- A red box appears in the aspect definition area of the Editor beneath the Description box.
- In the Name box in the Annotation Group area, enter the name of the group.
- Specify the fill color of the group rectangle that is used when a protein belongs to the new group as follows:
- Select Browse
next to Color to open the Color dialog box.
- Select the color that you want, or define a custom color. For information on defining a custom color, refer to the Set global default fragment match options topic in the Proteome Discoverer Help .
- Select OK.
- In the Annotation Database box, select the database of annotation to use.
- Enter the name that you are looking for in the filter cell.
- The entries in the table are automatically restricted to values containing the given name.
- To associate terms with the new group, select the terms to include in the list of available annotations, and then select the left arrow, , to move them to the Included Annotations pane.
- All terms are listed with an accession number and a description. The proteins in the results file reflect all terms, including the appended child terms, that you move to the group definition.
- For example, when you define a group with 17 different terms, the results report displays a colored rectangle if the corresponding protein is annotated with one of these 17 terms. Otherwise, the rectangle remains empty.