You can open the Annotation Aspects view from the Administration page.
Procedure
- Select Administration > Maintain Annotation Aspects.
- The Annotation Aspects view displays the currently available annotation aspects.
- Select the expand icon (+) to the left of an aspect.
- The application displays the GO Slim terms, annotation groups, or pathways associated with that aspect. It also displays the color code used in the results report for each group and the group name used in the ToolTip that appears when you move the cursor over the GO Slim category boxes.