If you have not yet restarted the server, you can undo any changes made in the FASTA Indexes Options dialog box since the last time that the server started.
Procedure
  1. Select Options.
  2. The FASTA Indexes Options dialog box opens.
  3. Select Reset.
  4. The changes that you made previously are deleted, and the directory reverts to its previous location.