You can restore the FASTA indexes listed under the Deleted FASTA Indexes table.
Procedure
- In the Deleted FASTA Indexes table, select the deleted index by selecting the arrow
to the left of the file name.
- Select Restore.
- In the Restore FASTA Indexes confirmation box, select OK.
- The restored index appears in the FASTA Indexes table and disappears from the Deleted FASTA Indexes table.