To view the result summaries, you must first open a result.
Prerequisites
- Open a result file.
Procedure
- From the application menu bar, select View > Summaries.
- Select the Configuration tab.
- On the right side of the Configuration page, select the Processing WF tab.
- The parameters are shown in the Configuration for Protein Scorer area.
- For information on the Result Summaries, see Using the Result Summaries.
Processing workflow configuration - To apply any changes to the settings of the Sequest protein-scoring configuration parameters, select Apply.
- When the message box opens, select OK.
NOTE
Select
to return to the default values.