Before using the Mascot node, set the default confidence threshold parameters.
Procedure
- From the application menu bar, select Administration > Configuration.
- Under Processing Settings, expand the Mascot folder.
- Select Mascot Node.
- The Default Confidence Thresholds (configuration parameters) for the Mascot node appear.
- Set the Default Confidence Thresholds parameters by doing one of the following:
- Select Significance High to calculate the thresholds for high-confidence peptides. The application automatically sets this value to the calculated relaxed significance when it performs a decoy search.
- Select Significance Middle to calculate the thresholds for medium-confidence peptides. The application automatically sets this value to the calculated relaxed significance when it performs a decoy search.
- Select Apply.
- Select OK in the Administration message box.
NOTE
Select
to return to the default values.