You can only use this procedure to update FASTA databases that you originally downloaded directly from the Annotation Server. The data on the Annotation Server is updated periodically.
Procedure
- Do one of the following:
- From the application menu bar, select Administration > Maintain FASTA Files.
- Select Maintain FASTA Files .
- The Administration page opens with the FASTA Files view.
- Select
to the left of a database row to check for updated information for that database.
- Select Check for Updates.
- The application now displays the search for updates as a job running in the job queue.
- When the job queue displays "Completed" in the Execution State column, close the Proteome Discoverer application and then reopen it.
- Select Administration > Maintain FASTA Files, or select Maintain FASTA Files .
- In the Update Available column of the FASTA Files view, the checkbox is selected for databases that have an available update.
- Select
to the left of a database row with the Update Available checkbox selected.
- The Update button becomes available.
- Select Update.
- The application displays the update as a job running in the job queue.
- (Optional) When the job queue displays "Completed" in the Execution State column, select FASTA Files under Content Management to return to the FASTA Files view.