You can import FASTA files from a local drive
Procedure
- Do one of the following:
- From the application menu bar, select Administration > Maintain FASTA Files.
- Select Maintain FASTA Files .
- The Administration page opens with the FASTA Files view.
- Select Add.
- In the Open dialog box, locate and select the FASTA file that you want to register, and then select Open.
- The FASTA file that you selected appears as a job in the job queue. To cancel the addition of this file, select Abort.
- When you see “Completed” in the Execution State column, the database has finished downloading.
- To add another FASTA file, do the following:
- Wait until the Execution State column indicates that the addition of the FASTA file is completed.
- Select FASTA Files in the left pane of the Administration page under Content Management.
- Select Add.
- The amount of time that it takes to import a FASTA file depends on the file size. When the application finishes importing a FASTA file, it displays “Available” in the Status column. This status indicates that the FASTA file is now available to use for a protein or peptide search with the Proteome Discoverer application.