Refine a filtered protein reference search - Refine a filtered protein reference search - Proteome Discoverer 3.1 User Guide - Proteome Discoverer Software - Refine a filtered protein reference search - Proteome Discoverer Software - Proteome Discoverer Software - topic
Proteome Discoverer 3.1 User Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Select the Custom option from the dropdown list below the operator.
- The Custom option opens the Custom Filter dialog box, so you can add multiple conditions.
Custom Filter dialog box- Select Add.
- A new line appears in the Operator (left) and Operand (right) lists.
- Select an operator from the Operator list.
- Enter an operand on the line in the Operand column.
- In the Filter Based On list, do one of the following:
- Select the All option to indicate whether the search algorithm should search for protein references that meet both conditions.
- Select the Any option to indicate whether the search algorithm should search for protein references that meet only one of the conditions.
- The following figure gives an example of a search for protein references that meet both of the conditions.
- Select OK.