When you or the Proteome Discoverer application deletes a FASTA file, it removes the FASTA indices belonging to the deleted FASTA file the next time that the server starts.
After you start the deletion of the FASTA file, you cannot cancel.
Procedure
- Do one of the following:
- From the application menu bar, select Administration > Maintain FASTA Files.
- Select Maintain FASTA Files .
- The Administration page opens with the FASTA Files view.
- Select the row of the FASTA file that you want to delete.
- Select Remove.
- In the Remove FASTA Databases dialog box, select OK.
- (Optional) In the Job Queue, remove the completed job for the selected FASTA file:
- Select Remove.
- In the Delete Jobs dialog box, select OK.