Before using the Mascot search engine, set the parameters that configure the Mascot server, the Mascot node, and the Mascot protein-scoring calculation.
Procedure
- From the application menu bar, select Administration > Configuration.
- Under Processing Settings, expand the Mascot folder.
- Select Mascot Server to open the parameters.
- The Proteome Discoverer application generates an MGF file that contains the search settings and all mass spectral information. It submits this file to the Mascot server through a web server, which might have a file size limitation. A search that generates large amounts of data—for example, a search with multiple raw data files—could create an MGF file that exceeds this limitation. The Max. MGF File Size [MB] parameter avoids this limitation by performing several separate Mascot searches and merging the results.
- Under Mascot Server, do the following:
- To split the MGF file and avoid any potential file-size limitations on the web server, enter the maximum size (in MB) that the MGF file can be in the Max. MGF File Size [MB] box.
NOTE
Make sure the size is less than the file size permitted by the web server. For information on these parameters, Mascot server configuration parameters</link>.
- In the Number of Attempts to Submit the Search box, specify the number of times that the application tries to submit the search when the Mascot server is busy.
- In the Time Interval Between Attempts to Submit a Search [sec] box, specify the interval of time, in seconds, that elapses between attempts to submit a search when the Mascot server is busy.
- When you are accessing a Mascot server through your own network and security for that server is enforced, enter your username and password in the corresponding boxes under Mascot Server Authentication.
- When you are accessing a Mascot server through the web and security for that server is enforced, enter your username and password in the corresponding boxes under Web Server Authentication.
- If you changed any settings of the Mascot server configuration parameters, select Apply.
- Select OK in the administration message box.
NOTE
Select
to return to the default values.