The Proteome Discoverer application generates many large temporary files—particularly when experiments involve label-free quantification and precursor ion quantification. To decrease wear on the C drive and to prevent filling the C drive with input raw data files and temporary results files, create a folder on another local drive to store the temporary files.
By default, the application stores temporary files in the directory:
C:\ProgramData\Thermo\Proteome Discoverer 3.1\Scratch
NOTE
If the C drive is smaller than 256 GB, create the Scratch folder in another drive.
If you create a folder with any name other than Scratch, the application creates a Scratch subfolder within your created folder.
Prerequisites
- You must have write access to the drive.
Procedure
- From the application menu bar, select Administration > Configuration.
- Under Server Settings, select Temporary Files.
- Select the browse button (...) next to the New Directory box.
- The Browse for Folder dialog box opens.
- Select the new location for the temporary files folder.
- Select OK.
- Select Apply.
- Restart the application.