The Start Page provides links to the most important commands that you use when working with the Proteome Discoverer application. It features the following three areas:
- Start, which contains the commands required to create a new study and analysis, open studies, and open results
- Recent Studies, which lists the 20 most recently opened studies
- Recent Results, which lists the 20 most recently opened result files
The links on the Start Page correspond to File menu commands.
Procedure
- To view the Start Page, do one of the following:
- On the Windows desktop, double-click Proteome Discoverer .
- Select View > Start Page.
- Select Start Page
from the application menu bar.
- The Start page opens.
- To open a result, see Open a result file.