If your processing computer is connected to the Internet, follow these instructions to deactivate the software license.
Procedure
- From the application menu bar, select Administration > Manage Licenses.
- On the License Manager page, select the software product that you want to deactivate, and then select Deactivate.
The License Deactivation dialog box opens. The Product ID box is populated with the selected product’s ID, and the Activation Code box is populated with the activation code for your license. - Select Online Deactivation.
- At the confirmation prompt, select Yes.
- The license deactivation process is complete, and the product ID disappears from the License Manager page.