Follow these instructions if your processing computer has an Internet connection.
Procedure
  1. From the application menu bar, select Administration > Manage Licenses.
  2. From the License Manager page, select Activate.
  3. The License Activation dialog box opens to the Activation Code view.
  4. Enter the licensing information.
  5. Select Online Activation to process the activation code.
  6. To close the dialog box, select OK.
This completes the online license activation process.