If you have files in other locations, the study management system automatically tries to locate files in a study folder or any of its subfolders.
Procedure
- Right-click the input file rows that contain invalid links.
- In the dropdown menu, select Find Missing Files.