You can move your study data to a new computer by copying a study folder with all of its data to another computer that has another installation of the Proteome Discoverer application.
Before you can move study data to a new computer, you must find any missing input files and update quantification methods. For more information, see Find missing input files and Update quantification methods.