You can export study definition files from an open study. It can be easier to export a study definition file from an existing study and modify it for a new dataset rather than creating a new file.
Procedure
  1. Open the study that requires a study definition file.
  2. Select File > Export > Study.
  3. Select a file name, and select OK.
  4. By default, the file name has the same name as the study and is saved as a tab-delimited text file.
  5. These exported files can be opened in Excel and saved as either a tab-delimited text file or an Excel file, which can both be imported into a new study.