Use the following procedure to filter multiple samples on the Samples page.
Procedure
- Select the Samples tab.
- Select the Contains filter
in the appropriate column of the Samples page, and select the type of filter from the list.
- In the box next to the icon, enter the text for the filter.
- For example, the following figure shows the results of changing the sample identifier to 126.
- Hold down the SHIFT key or CTRL key to highlight the rows to be modified.
- Right-click the highlighted cells, and select the study factor to apply.
- (Optional) To return the samples to their unfiltered state, select Clear All Filters
.