In this step, you specify the quantification method that was used for each of the files. In the example data set, all samples are labeled with the TMT 10plex corrected custom quantification method.
NOTE
You do not assign the quantification method to the input files for label-free quantification.
Procedure
  1. Select the Input Files tab.
  2. In each sample row, select the Quan Method column and select the quantification method (for this example, TMT 10plex) from the list.