You can add existing MSF files and result files to a study.
  • When you add an MSF file, the application also adds the input files to the study.
  • When you add a result file, the application adds the input files for each MSF file from the original result file to the study.
For information about reprocessing existing result files from other studies, see Reprocessing an existing analysis.