After you have added fractions to a study, you can optionally restrict data processing to a subset of the fractions. Use this approach to minimize data processing time during early workflow configuration or to remove a consistently bad fraction from a study.
Procedure
  1. In the study, select the Input Files tab.
  2. Select Show Details at the bottom left corner.
  3. Select Create Subset.
  4. The Create Subset dialog box opens.
  5. Select the checkboxes of the fractions that you want to include in the subset and then select Ok.
  6. Repeat taking fractions from each sample you want to analyze, select another sample and select Create Subset.
  7. In the Create Subset dialog box, select the fractions, and select Ok.
  8. The application creates new input file IDs that represent the subset of fractions.
  9. You can drag the new subset files into the Processing Step Files for Analysis in a new or existing analysis.