If the quantification method for the specific sample is not defined in the Proteome Discoverer application, see Add or change basic study information.
Procedure
  1. Select the Study Definition tab in the study.
  2. The Quantification Methods area of the Study Definition page lists all the quantification methods that have been imported into the study.
  3. To add a new Quantification Method to the study, select the appropriate quantification method for the current analysis from the Add dropdown list.
  4. After you add the Quantification Method to the study, you can assign the method to the raw data file. For information on adding input files, see Add Input Files.
  5. On the Study Definition page of the study, select the checkbox of the appropriate quantification method or methods in the Quantification Methods area.