On the Study Definition page, you can add or change some of the basic study information, which is also listed in the New Study and Analysis dialog box, as well as set up the study factors, which are not listed in the New Study and Analysis dialog box. You use study factors to describe and distinguish the quantification channels for your samples.
You can optionally add a description of the study by typing it in the Study Description area of the Study Definition page.