You can export the information from the Samples table to a text or Excel file for further edits. This feature will often be used as a template to create other studies.
Procedure
  1. Open the relevant study.
  2. Select File > Export > Study.
  3. In the Save As dialog, select either .txt or .xls in the Save as type dropdown.
  4. Select the appropriate target folder.
  5. Select a File name and select OK.
  6. By default, the file name will be the same name as the study.