You can copy and save views or graphs to various image formats.
Procedure
- To copy a view or a graph, open a view such as a Chromatogram Traces view (select View > Chromatogram Traces View).
- Right-click the view and select Copy.
- The application automatically copies the view as an image to the Clipboard. You can paste the image into another application.
- To save a view or graph in another format, open a view, such as a Chromatogram Traces view.
- Right-click the view and select Save As.
- Select the image type: EMF, PNG, GIF, JPG, TIFF, or BMP.
- (Optional) Select the location to store the image in.
- In the File Name box, enter the name of the file.
- Select Save.