You can export the protein references and sequences in your search results to a FASTA file.
Procedure
- Open your search results in the result file. See Organizing results.
- On the Proteins page, select the proteins that you want to export by selecting the checkbox next to the proteins that you want to export.
- Select File > Export > To FASTA.
- The Export to FASTA dialog box opens.
Export to FASTA dialog box - In the Destination Folder box, enter the path and name of the file where you want to save the exported protein references and sequences; or locate the file, select it, and select Save in the Save As dialog box.
- The application automatically changes the suffix of the results file from result to .fasta.
- To export only the references and sequences of the selected proteins to the FASTA file, do the following:
- In the Proteins page of the result file, select the checkbox to the left of each protein that you want to export. For information, see Check items on the pages of a result file.
- In the Export to FASTA dialog box, select the Checked Proteins Only checkbox.
- If you do not select the Checked Proteins Only checkbox, the application exports the references and sequences of all proteins.
- Select Export, or to close the Export to FASTA dialog box without exporting any results, select Close.