You can export spectra even though they do not identify any PSMs.
Procedure
- Open your result file. See Organizing results.
- On the MS/MS Spectrum Info page, sort the list by selecting the # PSMs column header.
- Select a checkbox in each row with a reported number of zero peptide matches.
- The result resembles the example in the following figure.
Selecting rows showing no PSM matches on the MS/MS Spectrum Info page - Select File > Export > Spectra.
- The Export Spectra dialog box opens.
Export Spectra dialog box - In the Destination box, enter the path and name of the file where you want to save the exported spectra; or locate and select the file, and then select Save in the Save As dialog box.
- Select the export format from the File Type list.
- In the Items To Be Exported area, select the From MS/MS Spectrum Info option and the Checked Items Only checkbox.
- In the Export Spectra dialog box, select Export.
- When you receive a message that the export job is finished, select Yes.