When you use the File > Export > Spectra command, you can select the spectra to export to a MGF, MZDATA, or MZML file.
For information on exporting spectra through a workflow, see Including Spectra Export in a Workflow.
Procedure
- Open your search results in the result file. See Organizing results.
- Select File > Export > Spectra.
- The Export Spectra dialog box opens.
- In the Destination Folder box, locate file where you want to save the exported spectra.
- In the File Type list, select the format of the file to export the spectra to:
- *.mgf: Mascot Generic Format (MGF) file
- *.mzData: MZDATA file
- *mzML: MZML file
- In the Items To Be Exported area of the Export Spectra dialog box, select the checkbox or boxes corresponding to the type of spectra that you want to export:
- From MS/MS Spectrum Info: Exports the spectra extracted from the spectrum file.
- From PSMs: Exports the spectra that identify the PSMs.
- From Peptide Groups: Exports the spectra that identify peptide groups. This option is available when the result file includes peptide groups.
- From Proteins: Exports the spectra that identify proteins. This option is available when the result file includes proteins.
- To export only certain spectra in the categories that you selected in the Items To Be Exported area, do the following:
- On the appropriate page of the result file, select the checkbox to the left of each item that you want to export.
- In the Items To Be Exported area of the Export Spectra dialog box, select the Checked Items Only checkbox.
- If you do not select the Checked Items Only checkbox, the application exports all spectra in the categories that you selected.
- Select Export, or to cancel this operation and any changes you made, select Close.
- When exporting is complete, you can open the exported file.