The application uses node-based workflows to process raw data files. Following set guidelines, you can create your own custom workflow nodes, or the Scripting Node provided with the application. Follow this procedure to install a scripting node.
Procedure
  1. Download the executable files and store them in the appropriate folder on the computer where you are running the application.
  2. Open the License Manager page.
  3. Select Scan for Missing Features to see third party nodes.
  4. Close and reopen the application.
  5. Select Help > About.
  6. The About Proteome Discoverer dialog box opens with the Patent and Legal Notices page displayed.
  7. Expand the Nodes list, and verify that it lists the new node.