In the node_response.json file, there are a number of options that can be selected for the new columns for specialized display in the result. The array Options holds these optional parameters as key-value pairs that are recognized when creating the new columns. There are different key values available:
PositionAfter—The new column will be placed after the specified column.PositionBefore—The new column will be placed before the specified column.RelativePosition—An integer value specifying the relative position of the column.PlotType- specifies the plot type where the new column can be plotted in a chart
- Valid values areNumeric,Categorical, andOrdinal. If there are multiple plot types, separate by comma (for example,Numeric, Categorical).
- If no plot type is defined:
• Boolean columns are automatically classified asCategorical
• Numeric columns are classified asNumeric, Ordinal.
• All others are specified asNone.FormatString- specified how numerical values are formatted and displayed. By default, float values have a format string of F5, which means they are displayed to 5 significant figures.SpecialCellRenderer—The data within a column can be shown with an alternative presentation rather than as a raw data value. Here are the possible options:
-exclamationMark—90550C2D-EB8D-443E-9F23-9E735FB23F9A - displays a string column. It shows an error icon if the value is not empty. The tool tip for the image is the full text.
-boolCheckMark—330D9522-50CC-41B7-9D45-5E5D8F708103 - displays a Boolean value. It shows a green check mark if the value is true.
-boolCrossMark—4B8956F1-3A54-423D-8654-60676825230F - displays a Boolean value. It shows a check mark (X) if the value is true.
-boolYesNo—9085D14E-388D-4EF0-91C1-B4B2BDF33AD3 - displays a Boolean value with the words "Yes" and "No".
-fileName—EB29D794-4F2E-4785-8B80-A24D8C0FB3E4 - for string column. Displays a filename (a string) by only showing the filename in the table field and showing the full path in the tooltip.
-fileSize—FBBFBB0B-A2D0-468F-95BF-25BCD0A42FE9 - displays an integer as a file size.
-percentBar—64516B23-565C-4036-87DA-29189E9E62A3 - displays a number between 0 and 100 as a column between 0% and 100%.
-monospaced—59E4C754-AD43-42FD-B4D9-C8CF0F622BD3 - displays any number or text using a monospaced font rather than a proportional font.
-concatenatedMultiline—410B767F-0C21-4E97-BB3D-041186955807 - displays a text that consists of multiple lines as one line that features the multiple lines concatenated using a ; (semicolon).