The Target Decoy PSM Validator node specifies the PSM confidences on the basis of dynamic score-based thresholds. It calculates the node-dependent score thresholds needed to determine the FDRs, which are given as input parameters of the node. The Target Decoy PSM Validator node needs results from a target and a decoy search.
The following table describes the parameters for the Target Decoy PSM Validator node.
Parameters | Description |
|---|---|
Target/Decoy Selection | Is used to toggle the FDR calculation separate or concatenated (default):
|
Target FDR (Strict) | Sets target false discovery rates for peptide matches of high confidence. Range: 0.0–1.0; default: 0.01 |
Target FDR (Relaxed) | Sets target false discovery rates for peptide matches of medium confidence. Range: 0.0–1.0; default: 0.05 |