The PSM Rescoring node is part of a predicted spectrum library workflow, where is in between the SequestHT node and the Percolator node. Using this node in the workflow, improves the confidence values on the Results page.
The INFERYS Rescoring node has no parameters.