The Comet version is 2019.01. For more information on Comet, visit https://uwpr.github.io/Comet/.
Comet node parameters
Parameters
Description
Protein Database
Specifies the sequence database to be searched.
Comet Params File
The parameter file for Comet. Leave this empty to use the default parameters for the installed version of Comet. Insert the text of a custom parameter file if any changes should be used.
Note Modifications should be set by the modification parameters below. Modifications in the given text may be overwritten.
Variable Modifications
Select any known or suspected modification.
Variable Modifications (peptide terminus)
N-Terminal Modification
Select any known or suspected N-terminal modification. Dynamic modifications are those which may or may not be present.
C-Terminal Modification
Select any known or suspected C-terminal modification. Dynamic modifications are those which may or may not be present.
Peptide N-Terminus
Static modification for the N-terminus of the peptide used during the search. Static modifications are applied universally to every instance of the specified residue(s) or terminus.
Peptide C-Terminus
Static modification for the C-terminus of the peptide used during the search. Static modifications are applied universally to every instance of the specified residue(s) or terminus.
Static Modification
Select any known or suspected modification. Static modifications are applied universally to every instance of the specified residue(s) or terminus.