The Mascot node analyzes the data using the Mascot search engine.
The following table describes the parameters for the Mascot node.
Parameter | Definition |
|---|---|
Input Data |
|
Protein Database | Specifies the sequence databases to use in the search. The available databases are configured on the Mascot server. If you use Mascot server 2.3 or later, you can use more than one sequence database for the same search. |
Enzyme Name | Specifies the enzyme reagent used for protein digestion. The following enzymes, set up by the Mascot server, are typical of what might be available:
|
Maximum Missed Cleavage Sites | Specifies the maximum number of missed cleavage sites to consider during the digest. Range: 0 – 9; default: 1 |
Instrument | Specifies the type of instrument used to acquire the data in the raw data file:
Note The available instruments might vary on your Mascot server. |
Taxonomy | Limits searches to entries from a particular species or group of species. Can speed up a search and ensure that only the match list contains entries from the selected species. Select from the taxonomy database list. |
User Email | Specifies the user email address sent to Mascot for a search. |
Tolerances |
|
Precursor Mass Tolerance | Specifies the mass tolerance used for finding peptide candidates. You can specify the units (Da, mmu, or ppm) for the precursor mass tolerance value. Range: 0.0001–5.0 Da or 0.04–5000 ppm; default: 10 ppm |
Fragment Mass Tolerance | Specifies the mass tolerance used for matching fragment peaks. You can specify the units (Da or mmu) for the fragment mass tolerance value. Range: 0.0001 Da–2.0 Da; default: 0.8 Da |
Use Average Precursor Mass | Determines the type of mass to use for matching the precursor.
|
Modification Groups |
|
From Quan Method | Uses the modifications specified within the modification group of the selected quantification method in addition to the specified dynamic and static modifications. You can select these modifications from a dropdown list. Do not use the modifications that you specify in the modification groups in the selected quantification method as additional dynamic or static modifications. For more information on Mascot quantification methods, see Checking the quantification method. |
Dynamic Modifications |
|
Show All Modifications | Determines whether to display a short list of the most common modifications or a complete list of all available modifications.
|
Dynamic Modification | Determines PTMs. The Mascot administrator defines the available modifications. The application modifies some amino acid residues but not others. |
Static Modifications |
|
Static Modification | Applies the same specific mass to all occurrences of the named amino acid, as in an exhaustive chemical modification. The Mascot administrator defines the available modifications. The application applies static modifications universally to every instance on the specified residues or terminus. |