The Reporter Ions Quantifier node performs reporter ion quantification, which uses external reagents, or tags, to enzymatically or chemically label proteins and peptides. The application uses this node in iTRAQ and TMT experiments.
For more information on using the node in a workflow, see Creating Reporter Ion Quantification Workflows.
The following table describes the parameters for the Reporter Ions Quantifier node.
Parameter | Definition |
|---|---|
Integration Tolerance | Specifies the mass-to-charge window so that you can look for the reporter peaks. Range: 0.0001–0.6 Da or 0.01–5000 ppm; default: 20 ppm |
Integration Method | Specifies which peak to select if more than one peak is found inside the integration window:
|
Mass Analyzer | Specifies the type of mass spectrometer used in the acquisition of the spectrum:
|
MS Order | Specifies the level of the mass spectrum to be processed, for example, MS/MS or MS3. Range: MS2–MS10; default: MS2 |
Activation Type | Specifies the fragmentation method used for each spectrum:
For descriptions, see Multidimensional separation experiments. |
Min. Collision Energy | Filters out fragment spectra with a normalized collision energy below the specified value. Range: 0–100; default: 0 |
Max. Collision Energy | Filters out fragment spectra with a normalized collision energy above the specified value. Range: 0–1000; default: 1000 |