All processing workflows start with a spectrum files node, which submits the selected input files to the downstream nodes in the workflow. The Data Input area of the Processing Workflow window contains two spectrum files nodes.