The quantification spectra on the pages of the result file show the isotope pattern used for quantifying the peptides. The algorithm used in precursor ion quantification finds isotope patterns by identifying target components—that is, known elemental compositions from event lists. It identifies the peptides and searches in the event lists for the isotope patterns of these identified peptides. After peptide identification, the algorithm follows the steps shown in the following figure to identify the isotope patterns.